Orders LinkContactUsLinkComponent
Retort rings
Retort rings
Retort rings
Certificates
10 Options Available Below
Product Details & Documents
Steel, epoxy coated.
  • Available with or without bosshead
  • Bosshead is made of malleable iron
Product Family Options
Without bosshead
Product Information
  • Ext.Ø
    Availability
    Price
  • JUCHVL04890
    Ring without bosshead, 107 g
    Ring without bosshead, 107 g
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • JUCHVL04900
    Ring without bosshead, 157 g
    Ring without bosshead, 157 g
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • JUCHVL04920
    Ring without bosshead, 230 g
    Ring without bosshead, 230 g
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 18,60 €
    18,60 €
    1 * 1 items
       18,60 €
  • JUCHVL04930
    Ring without bosshead, 260 g
    Ring without bosshead, 260 g
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 19,50 €
    19,50 €
    1 * 1 items
       19,50 €
  • JUCHVL04932
    Ring without bosshead, 346 g
    Ring without bosshead, 346 g
    180 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
With bosshead
Product Information
  • Ext.Ø
    Availability
    Price
  • JUCHVL04940
    Ring with bosshead, 165 g
    Ring with bosshead, 165 g
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 19,70 €
    19,70 €
    1 * 1 items
       19,70 €
  • JUCHVL04950
    Ring with bosshead, 210 g
    Ring with bosshead, 210 g
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 22,40 €
    22,40 €
    1 * 1 items
       22,40 €
  • JUCHVL04960
    Ring with bosshead, 236 g
    Ring with bosshead, 236 g
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 24,90 €
    24,90 €
    1 * 1 items
       24,90 €
  • JUCHVL04970
    Ring with bosshead, 266 g
    Ring with bosshead, 266 g
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • JUCHVL04980
    Ring with bosshead, 290 g
    Ring with bosshead, 290 g
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items