alpha-Amylase (from Bacillus licheniformis)
Catalog # SIALA3306-10ML
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mL
- CAS No.:9000-85-5
- Source:Bacillus licheniformis
- Species:
- Environmentally Preferable:
- Form:Solution
- Enzyme name:alpha-Amylase
Specifications
About this item
α-Amylase solution is used in total dietary fiber assay.
- Heat stable
- Store at 2 to 8 °C
Enhanced for waste prevention when used in starch ethanol research.