Order Entry
Export
ContactUsLinkComponent
Cortisol express ELISA kit
Cortisol express ELISA kit
Supplier:  Cayman Chemical
Certificates
undefined
Cortisol express ELISA kit
Supplier:  Cayman Chemical

Specifications

  • Developmental time:
    120 min
  • Mid point:
    300 - 500 pg/ml (50% B/B₀)
  • Detection range:
    39,1 - 5000 pg/ml
  • Storage temperature:
    –20 °C
  • Sensitivity:
    110 pg/ml (80% B/B₀)
  • Incubation time:
    2 hr
  • Assay Type:
    Competitive
  • Format:
    Pre-coated
  • Target protein:
    Cortisol

Specifications

Related Information