Orders LinkContactUsLinkComponent
Disposable cuvettes, round
Disposable cuvettes, round
Disposable round cuvettes, glass, without cap, 11 mm
Catalog # HACHLZM137
Supplier:   Hach
Disposable cuvettes, round
Catalog # HACHLZM137
Supplier:  Hach
Disposable cuvettes, round
Disposable round cuvettes, glass, without cap, 11 mm
Price
1 910,00 €
Log in for availability
1 * 560 items
Product Details & Documents
These round cuvettes are made of glass.
Ordering information: For compatible screw caps, see series HACHLZM383.
Specifications
  • Description:
    Disposable round cuvettes, glass, without cap
  • Height:
    11 mm