Order Entry
Export
ContactUsLinkComponent
ASTM® D3606 backflush standard (2,2,4-trimethylpentane (isooctane))
ASTM® D3606 backflush standard (2,2,4-trimethylpentane (isooctane))
Catalog # REST30671
Supplier:  Restek
CAS Number:  
undefined
ASTM® D3606 backflush standard (2,2,4-trimethylpentane (isooctane))
Catalog # REST30671
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    5% vol/vol in nonane
  • Pk:
    1 mL
  • Description:
    ASTM D3606 backflush standard (2,2,4-trimethylpentane (isooctane))
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.