Creosote oil standard
REST31838
: Restek
:
- Concentration:50000 µg/ml in methylene chloride
- Pk:1 Ampoul
- Description:Creosote oil standard
- Environmentally Preferable:
- Pk Info:1 ml/ampoule
This is for total petroleum hydrocarbon pattern recognition of creosote oil.
Creosote oil, a widely used wood preservative produced by distilling coal tar, contains chemicals that are classified as carcinogens (benzo(a)pyrene).