ASTM D3606 calibration standard #7 without internal standard (2 components)
Catalog # REST30653
Supplier: Restek
CAS Number:
Specifications
- Pk:1 Ampoul
- Description:ASTM D3606 calibration standard #7 without internal standard, in isooctane
- Pk Info:25 ml/ampoule
Specifications
About this item
Contains two components, those are benzene and toluene.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.