Order Entry
Export
ContactUsLinkComponent
ASTM D3606 calibration standard #6 without internal standard (2 components)
ASTM D3606 calibration standard #6 without internal standard (2 components)
Catalog # REST30652
Supplier:  Restek
CAS Number:  
undefined
ASTM D3606 calibration standard #6 without internal standard (2 components)
Catalog # REST30652
Supplier:  Restek
CAS Number:  

Specifications

  • Pk:
    1 Ampoul
  • Description:
    ASTM D3606 calibration standard #6 without internal standard, in isooctane
  • Pk Info:
    25 ml/ampoule

Specifications

About this item

Contains two components, those are benzene and toluene.

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.