Hexachlorophene
Hexachlorophene, 2000 μg/ml in methylene chloride, 1 ml/ampoule, 1 Ampoul
Catalog # REST31811
Supplier: Restek
Jump to:
Product Details & Documents
: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Specifications
Specifications
- Concentration:2000 μg/ml in methylene chloride
- Pk:1 Ampoul
- Description:Hexachlorophene
- Pk Info:1 ml/ampoule
- Cat. No.:REST31811
- Formula:C₁₃H₆Cl₆O₂
- Description:Hexachlorophene
- CAS No.:70-30-4



