Order Entry
Export
Orders LinkContactUsLinkComponent
Benzo(a)pyrene

Benzo(a)pyrene

Benzo(a)pyrene, 1000 μg/ml in acetone, 1 ml/ampoule, 1 mL
Catalog # REST31271
Supplier:   Restek
Benzo(a)pyrene
Catalog # REST31271
Supplier:  Restek
CAS Number:  
Benzo(a)pyrene
Benzo(a)pyrene

Benzo(a)pyrene, 1000 μg/ml in acetone, 1 ml/ampoule, 1 mL

CAS Number:  
Price
54,50 €
Log in for availability
1 * 1 mL
Product Details & Documents

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.

Specifications

Specifications

  • Concentration:
    1000 μg/ml in acetone
  • Pk:
    1 mL
  • Description:
    Benzo(a)pyrene
  • Pk Info:
    1 ml/ampoule
  • Cat. No.:
    REST31271
  • Formula:
    C₂₀H₁₂
  • CAS No.:
    50-32-8

Specifications