Acenaphthylene
Acenaphthylene, 1000 μg/mL in methanol, 1 ml/ampoule, 1 mL
REST31268
: Restek
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
- Concentration:1000 μg/mL in methanol
- Pk:1 mL
- Description:Acenaphthylene
- Pk Info:1 ml/ampoule
- Cat. No.:REST31268
- Formula:C₁₂H₈
- CAS No.:208-96-8



