FAME #18 mix (10 components)
Catalog # REST35024
Supplier: Restek
CAS Number:
Specifications
- Concentration:Neat
- Pk:1 mL
- Description:FAME #18 mix
- Pk Info:50 mg/ampoule
Specifications
About this item
This mixture can be used for quantification (AOCS Method CE 1a–13) and approximate the compositions of the types of oils like peanut, rapeseed and mustard seed oil.
This mix contains ten components such as C14:00 methyl myristate (1%), C16:00 methyl palmitate (4%), C18:00 methyl stearate (3%), C18:01 methyl oleate (45%), C18:02 methyl linoleate (15%), C18:03 methyl linolenate (3%), C20:00 methyl arachidate (3%), C22:00 methyl behenate (3%), C22:01 methyl erucate (20%), C24:00:00 methyl lignocerate (3%).
% represents weight/weight% basis (minimum 50 mg/ampoule).