Dinoseb
Dinoseb, 1000 µg/ml in methanol, 1 ml/ampoule, 1
Catalog # REST32251
Supplier: Restek
Jump to:
Product Details & Documents
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Specifications
Specifications
- Concentration:1000 µg/ml in methanol
- Pk:1
- Description:Dinoseb
- Pk Info:1 ml/ampoule
- Cat. No.:REST32251
- Formula:C₁₀H₁₂N₂O₅
- CAS No.:88-85-7



