Order Entry
Export
Orders LinkContactUsLinkComponent
Hexachlorobenzene

Hexachlorobenzene

Hexachlorobenzene, 1000 µg/ml in acetone, 1 ml/ampoule, 1
Catalog # REST32231
Supplier:   Restek
Hexachlorobenzene
Catalog # REST32231
Supplier:  Restek
CAS Number:  
Hexachlorobenzene
Hexachlorobenzene

Hexachlorobenzene, 1000 µg/ml in acetone, 1 ml/ampoule, 1

CAS Number:  
Price
48,50 €
Log in for availability
1 * 1 items
Product Details & Documents

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.

Specifications

Specifications

  • Concentration:
    1000 µg/ml in acetone
  • Pk:
    1
  • Description:
    Hexachlorobenzene
  • Pk Info:
    1 ml/ampoule
  • Cat. No.:
    REST32231
  • Formula:
    C₆Cl₆
  • CAS No.:
    118-74-1

Specifications