Order Entry
Export
Orders LinkContactUsLinkComponent
4,4'-DDT

4,4'-DDT

4,4'-DDT, 1000 µg/ml in methanol, 1 ml/ampoule, 1
Catalog # REST32203
Supplier:   Restek
4,4'-DDT
Catalog # REST32203
Supplier:  Restek
CAS Number:  
4,4'-DDT
4,4'-DDT

4,4'-DDT, 1000 µg/ml in methanol, 1 ml/ampoule, 1

CAS Number:  
Price
53,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.

Specifications

Specifications

  • Concentration:
    1000 µg/ml in methanol
  • Pk:
    1
  • Description:
    4,4'-DDT
  • Pk Info:
    1 ml/ampoule
  • Cat. No.:
    REST32203
  • Formula:
    C₁₄H₉Cl₅
  • CAS No.:
    50-29-3

Specifications