Order Entry
Export
Orders LinkContactUsLinkComponent
Kovats probes test standard (4 components)

Kovats probes test standard (4 components)

Kovats probes test standard, 500 µg/ml each in methylene chloride, 1 ml/ampoule, 1
Catalog # REST35259
Supplier:   Restek
Kovats probes test standard (4 components)
Catalog # REST35259
Supplier:  Restek
CAS Number:  
Kovats probes test standard (4 components)
Kovats probes test standard (4 components)

Kovats probes test standard, 500 µg/ml each in methylene chloride, 1 ml/ampoule, 1

CAS Number:  
Price
52,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Contains: Benzene, 1-butanol, 1-nitropropane, 2-pentanone.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.

Specifications

Specifications

  • Concentration:
    500 µg/ml each in methylene chloride
  • Pk:
    1
  • Description:
    Kovats probes test standard
  • Pk Info:
    1 ml/ampoule

Specifications