Order Entry
Export
Orders LinkContactUsLinkComponent
 

WA VPH standard (15 components)

WA VPH standard, 1000 μg/ml in P and T methanol, 1 ml/ampoule, 1 mL
Catalog # REST30451
Supplier:   Restek
WA VPH standard (15 components)
Catalog # REST30451
Supplier:  Restek
CAS Number:  
WA VPH standard (15 components)
WA VPH standard (15 components)

WA VPH standard, 1000 μg/ml in P and T methanol, 1 ml/ampoule, 1 mL

CAS Number:  
Price
85,50 €
Log in for availability
1 * 1 mL
Product Details & Documents

WA VPH standard contains (C5) n-pentane, (C6) n-hexane, (C8) n-octane, (C10) n-decane, (C12) n-dodecane
benzene, ethylbenzene, 1-methylnaphthalene, methyl tert-butyl ether (MTBE), naphthalene, toluene, 1,2,3-trimethylbenzene, m-xylene, o-xylene, p-xylene.

 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.

Specifications

Specifications

  • Concentration:
    1000 μg/ml in P and T methanol
  • Pk:
    1 mL
  • Description:
    WA VPH standard
  • Pk Info:
    1 ml/ampoule

Specifications