Order Entry
Export
Orders LinkContactUsLinkComponent
Grob mix 280 (10 Components)

Grob mix 280 (10 Components)

Grob mixture 280, In methylene chloride, 1 ml/ampoule, 1 Ampoul
Catalog # REST35227
Supplier:   Restek
Grob mix 280 (10 Components)
Catalog # REST35227
Supplier:  Restek
CAS Number:  
Grob mix 280 (10 Components)
Grob mix 280 (10 Components)

Grob mixture 280, In methylene chloride, 1 ml/ampoule, 1 Ampoul

CAS Number:  
Price
63,50 €
Log in for availability
1 * 1 Ampoul
Product Details & Documents

The grob mix 280 consists of 10 components n-decane (280 μg/ml), methyl decanoate (420 μg/ml), n-undecane (280 μg/ml), methyl undecanoate (420 μg/ml), methyl dodecanoate (420 μg/ml), L-2,3-butanediol (530 μg/ml), 2,6-dimethylaniline (320 μg/ml), 2,6-dimethylphenol (320 μg/ml), nonanal (400 μg/ml) and 1-octanol (360 μg/ml).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.

Specifications

Specifications

  • Concentration:
    In methylene chloride
  • Pk:
    1 Ampoul
  • Description:
    Grob mixture 280
  • Pk Info:
    1 ml/ampoule

Specifications