N-nitrosodimethylamine-d6
N-nitrosodimethylamine-d6, 1000 µg/ml in methylene chloride, 1 ml/ampoule, 1 mL
Catalog # REST33910
Supplier: Restek
Jump to:
Product Details & Documents
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Specifications
Specifications
- Concentration:1000 µg/ml in methylene chloride
- Pk:1 mL
- Description:N-nitrosodimethylamine-d6
- Pk Info:1 ml/ampoule
- Cat. No.:REST33910
- Formula:C₂H₆N₂O
- Description:N-Nitrosodimethylamine-d6
- CAS No.:17829-05-9



