Order Entry
Export
ContactUsLinkComponent
Internal standard, method 527 (3 components)
Internal standard, method 527 (3 components)
Catalog # REST33010
Supplier:  Restek
CAS Number:  
undefined
Internal standard, method 527 (3 components)
Catalog # REST33010
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    500 µg/ml each in acetone
  • Pk:
    1 Ampoul
  • Description:
    Internal standard, method 527
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The internal standard consists of three components acenaphthene-d10, chrysene-d12 and phenanthrene-d10.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.