Order Entry
Export
ContactUsLinkComponent
Motor oil composite standard
Motor oil composite standard
Catalog # REST31464
Supplier:  Restek
CAS Number:  
undefined
Motor oil composite standard
Catalog # REST31464
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    50000 μg/ml in methylene chloride
  • Pk:
    1 mL
  • Description:
    Motor oil composite
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.