Order Entry
Export
Orders LinkContactUsLinkComponent
Styrene

Styrene

Styrene, 2000 μg/ml in P and T methanol, 1 ml/ampoule, 1 mL
Catalog # REST30410
Supplier:   Restek
Styrene
Catalog # REST30410
Supplier:  Restek
CAS Number:  
Styrene
Styrene

Styrene, 2000 μg/ml in P and T methanol, 1 ml/ampoule, 1 mL

CAS Number:  
Price
52,50 €
Log in for availability
1 * 1 mL
Product Details & Documents

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.

Specifications

Specifications

  • Concentration:
    2000 μg/ml in P and T methanol
  • Pk:
    1 mL
  • Description:
    Styrene
  • Pk Info:
    1 ml/ampoule
  • Cat. No.:
    REST30410
  • Formula:
    C₈H₈
  • CAS No.:
    100-42-5

Specifications