2,2-Dichloropropane
REST30283
: Restek
:
- Concentration:2000 μg/ml in P and T methanol
- Pk:1
- CAS No.:594-20-7
- Description:2,2-Dichloropropane
- Formula:C₃H₆Cl₂
- Pk Info:1 ml/ampoule
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.