Trans-1,4-dichloro-2-butene
Catalog # REST30274
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml in P&T methanol
- Pk:1 mL
- CAS No.:64742-65-0
- Description:trans-1,4-Dichloro-2-butene
- Formula:C₄H₆Cl₂
- Pk Info:1 ml/ampoule
Specifications
About this item
Prepared from an equal-volume blend of 5W30, 10W30, 10W40, and 20W50 motor oils. After blending, a precisely weighed amount of the composite is added to a volumetric flask to produce the standard.
Motor Oil standards may exhibit lot-to-lot variation.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.