1,2-Dibromo-3-chloropropane
REST30270
: Restek
:
- Concentration:2000 μg/ml in P and T methanol
- Pk:1
- CAS No.:96-12-8
- Description:1,2-Dibromo-3-chloropropane (DBCP)
- Formula:C₃H₅Br₂Cl
- Pk Info:1 ml/ampoule
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.