2-Chloroethanol
2-Chloroethanol, 2000 μg/ml in P and T methanol, 1 ml/ampoule, 1
REST30264
: Restek
Jump to:
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifications
Specifications
- Concentration:2000 μg/ml in P and T methanol
- Pk:1
- Description:2-Chloroethanol
- Pk Info:1 ml/ampoule
- Cat. No.:REST30264
- Formula:C₂H₅ClO
- CAS No.:107-07-3



