Order Entry
Export
Orders LinkContactUsLinkComponent
Bis(2-ethylhexyl)adipate

Bis(2-ethylhexyl)adipate

Bis(2-ethylhexyl)adipate, 1000 µg/ml in methanol, 1 ml/ampoule, 1 mL
Catalog # REST31449
Supplier:   Restek
Bis(2-ethylhexyl)adipate
Catalog # REST31449
Supplier:  Restek
CAS Number:  
Bis(2-ethylhexyl)adipate
Bis(2-ethylhexyl)adipate

Bis(2-ethylhexyl)adipate, 1000 µg/ml in methanol, 1 ml/ampoule, 1 mL

CAS Number:  
Price
40,00 €
Log in for availability
1 * 1 mL
Product Details & Documents

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.

Specifications

Specifications

  • Concentration:
    1000 µg/ml in methanol
  • Pk:
    1 mL
  • Description:
    Bis(2-ethylhexyl)adipate
  • Pk Info:
    1 ml/ampoule
  • Cat. No.:
    REST31449
  • Formula:
    C₂₂H₄₂O₄
  • Description:
    Bis(2-ethylhexyl)adipate
  • CAS No.:
    103-23-1

Specifications