1,2-Dichlorobenzene
1,2-Dichlorobenzene, 1000 µg/m in methanol, 1 ml/ampoule, 1 mL
Catalog # REST31442
Supplier: Restek
Jump to:
Product Details & Documents
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifications
Specifications
- Concentration:1000 µg/m in methanol
- Pk:1 mL
- Description:1,2-Dichlorobenzene
- Pk Info:1 ml/ampoule
- Cat. No.:REST31442
- Formula:C₆H₄Cl₂
- CAS No.:95-50-1



