Bis(2-ethylhexyl)phthalate
Bis(2-ethylhexyl)phthalate, 1000 µg/ml in methylene chloride, 1 ml/ampoule, 1 mL
Catalog # REST31420
Supplier: Restek
Jump to:
Product Details & Documents
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifications
Specifications
- Concentration:1000 µg/ml in methylene chloride
- Pk:1 mL
- Description:Bis(2-ethylhexyl)phthalate
- Pk Info:1 ml/ampoule
- Cat. No.:REST31420
- Formula:C₂₄H₃₈O₄
- CAS No.:117-81-7



