Order Entry
Export
ContactUsLinkComponent
DHA PiONA standard (133 components)
DHA PiONA standard (133 components)
Catalog # REST30730
Supplier:  Restek
CAS Number:  
undefined
DHA PiONA standard (133 components)
Catalog # REST30730
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Pk:
    1
  • Description:
    DHA PiONA standard
  • Pk Info:
    0,15 ml in autosampler vial

Specifications

About this item

This standard is produced for refineries performing Detailed Hydrocarbon Analysis (DHA) of crude oil feedstocks and fuels.

It contains 133 components.

It helps with calibrating complex hydrocarbon analyses and provides the greatest number of gravimetrically determined values for quantitative calibration.

Certifications: ASTM Methods D6729, D6730, and D6733 (Determination of Individual Components in Spark Ignition Engine Fuels)