UST Diesel surrogate standard
Catalog # REST31098
Supplier: Restek
CAS Number:
Specifications
- Concentration:10000 μg/ml in methylene chloride
- Pk:1
- Description:UST Diesel surrogate standard
- Pk Info:1 ml/ampoule
Specifications
About this item
Contains 1-chlorooctadecane.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Delivery: Includes an ampoule breaker, deactivated amber screw-top vial, extra product label, certificate of analysis, and material safety data sheet (MSDS).