Order Entry
Export
ContactUsLinkComponent
DHA aromatics standard (37 components)
DHA aromatics standard (37 components)
Catalog # REST30729
Supplier:  Restek
CAS Number:  
undefined
DHA aromatics standard (37 components)
Catalog # REST30729
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Pk:
    0,15 mL
  • Description:
    DHA Aromatics Standard
  • Pk Info:
    0,15 ml in autosampler vial

Specifications

About this item

This standard is produced for refineries performing Detailed Hydrocarbon Analysis (DHA) of crude oil feedstocks and fuels.

This standard contain 37 components: Benzene (8,255%), N-Butylbenzene (2,077%), sec-Butylbenzene (3,09%), tert-Butylbenzene (2,058%), tert-1-Butyl-4-Ethylbenzene (2,076%), tert-1-Butyl-2-Methylbenzene (2,077%), tert-1-Butyl-3,4,5-Trimethylbenzene (3,355%), 1,2-Dimethyl-3-Ethylbenzene (2,152%), 1,2-Dimethyl-4-Ethylbenzene (0,915%), 1,3-Dimethyl-2-Ethylbenzene (0,799%), 1,3-Dimethyl-5-Ethylbenzene (0,407%), 1,4-Dimethyl-2-Ethylbenzene (2,076%), Ethylbenzene (8,248%), Hexylbenzene (5,155%), Isobutylbenzene (3,099%), Isopropylbenzene (2,085%), 2-Methylbutylbenzene (0,118%).

It helps with calibrating complex hydrocarbon analyses and provides the greatest number of gravimetrically determined values for quantitative calibration.

Certifications: Meets requirements such as ASTM Methods D6729, D6730, and D6733. Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.