Order Entry
Export
ContactUsLinkComponent
Herbicide reference standard, 2,4-D methyl ester
Herbicide reference standard, 2,4-D methyl ester
Catalog # REST32240
Supplier:  Restek
CAS Number:  
undefined
Herbicide reference standard, 2,4-D methyl ester
Catalog # REST32240
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    1,000 µg/ml in methanol
  • Pk:
    1
  • CAS No.:
    1928-38-7
  • Description:
    2,4-D methyl ester
  • Formula:
    C₉H₈Cl₂O₃
  • Description:
    2,4-D methyl ester
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.