Order Entry
Export
Orders LinkContactUsLinkComponent
 

Atlantic RBCA EPH mix (11 components)

Atlantic RBCA VPH mix, 1000 µg/ml each in hexane:methylene chloride (85:15%), 1 ml/ampoule, 1 Ampoul
Catalog # REST31872
Supplier:   Restek
Atlantic RBCA EPH mix (11 components)
Catalog # REST31872
Supplier:  Restek
CAS Number:  
Atlantic RBCA EPH mix (11 components)
Atlantic RBCA EPH mix (11 components)

Atlantic RBCA VPH mix, 1000 µg/ml each in hexane:methylene chloride (85:15%), 1 ml/ampoule, 1 Ampoul

CAS Number:  
Price
58,00 €
Log in for availability
1 * 1 Ampoul
Product Details & Documents

Contains: n-decane, C10, n-dodecane, C12, n-hexadecane, C16, n-heneicosane, C21, n-octacosane, C28, n-dotriacontane, C32, acenaphthene, anthracene, benzo(a)pyrene, chrysene, naphthalene.

 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.

Specifications

Specifications

  • Concentration:
    1000 µg/ml each in hexane:methylene chloride (85:15%)
  • Pk:
    1 Ampoul
  • Description:
    Atlantic RBCA VPH mix
  • Pk Info:
    1 ml/ampoule

Specifications