Orders LinkContactUsLinkComponent
Oxygenates (6 components)
Oxygenates, in P&T methanol, 1 ml/ampoule, 1 Ampoul
Catalog # REST30626
Supplier:   Restek
Oxygenates (6 components)
Catalog # REST30626
Supplier:  Restek
CAS Number:  
Oxygenates (6 components)
Oxygenates (6 components)
Oxygenates, in P&T methanol, 1 ml/ampoule, 1 Ampoul
CAS Number:  
Price
74,00 €
Log in for availability
1 * 1 Ampoul
Product Details & Documents
This Oxygenates compound includes six components: tert-amyl ethyl ether (TAEE) 2000 μg/ml, tert-Amyl methyl ether (TAME) 2000 μg/ml, tert-Butanol (TBA) 10000 μg/ml, Diisopropyl ether (DIPE) 2000 μg/ml, Ethyl-tert-butyl ether (ETBE) 2000 μg/ml and Methyl tert-butyl ether (MTBE) 2000 μg/ml.
 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifications
Specifications
  • Pk:
    1 Ampoul
  • Description:
    Oxygenates, in P&T methanol
  • Pk Info:
    1 ml/ampoule
Specifications