Order Entry
Export
Orders LinkContactUsLinkComponent
Acetophenone

Acetophenone

Acetophenone, 5000 µg/ml in P&T methanol, 1 ml/ampoule, 1 Ampoul
Catalog # REST30621
Supplier:   Restek
Acetophenone
Catalog # REST30621
Supplier:  Restek
CAS Number:  
Acetophenone
Acetophenone

Acetophenone, 5000 µg/ml in P&T methanol, 1 ml/ampoule, 1 Ampoul

CAS Number:  
Price
48,00 €
Log in for availability
1 * 1 Ampoul
Product Details & Documents

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.

Specifications

Specifications

  • Concentration:
    5000 µg/ml in P&T methanol
  • Pk:
    1 Ampoul
  • Description:
    Acetophenone
  • Pk Info:
    1 ml/ampoule
  • Cat. No.:
    REST30621
  • Formula:
    C₈H₈O
  • Description:
    Acetophenone
  • CAS No.:
    98-86-2

Specifications