Phenanthrene
Phenanthrene, 1000 µg/ml in methanol, 1 ml/ampoule, 1 mL
REST31281
: Restek
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
- Concentration:1000 µg/ml in methanol
- Pk:1 mL
- Description:Phenanthrene
- Pk Info:1 ml/ampoule
- Cat. No.:REST31281
- Formula:C₁₄H₁₀
- Description:Phenanthrene
- CAS No.:85-01-8



