Naphthalene
Naphthalene, 1000 µg/ml in methanol, 1 ml/ampoule, 1 mL
Catalog # REST31280
Supplier: Restek
Jump to:
Product Details & Documents
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifications
Specifications
- Concentration:1000 µg/ml in methanol
- Pk:1 mL
- Description:Naphthalene
- Pk Info:1 ml/ampoule
- Cat. No.:REST31280
- Formula:C₁₀H₈
- Description:Naphthalene
- CAS No.:91-20-3



