Order Entry
Export
Orders LinkContactUsLinkComponent
 

Chrysene

Chrysene, 1000 µg/ml in acetone, 1 ml/ampoule, 1 mL
  REST31275
 :   Restek
Chrysene
  REST31275
 :  Restek
 :  
Chrysene
Chrysene

Chrysene, 1000 µg/ml in acetone, 1 ml/ampoule, 1 mL

 :  
 
56,50 €
   
1 * 1 mL
 

 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.

 

 

  • Concentration:
    1000 µg/ml in acetone
  • Pk:
    1 mL
  • Description:
    Chrysene
  • Pk Info:
    1 ml/ampoule
  • Cat. No.:
    REST31275
  • Formula:
    C₁₈H₁₂
  • Description:
    Chrysene
  • CAS No.:
    218-01-9