Benz(a)anthracene
Benz(a)anthracene, 1000 µg/ml in methanol, 1 ml/ampoule, 1 mL
REST31270
: Restek
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
- Concentration:1000 µg/ml in methanol
- Pk:1 mL
- Description:Benz(a)anthracene
- Pk Info:1 ml/ampoule
- Cat. No.:REST31270
- Formula:C₁₈H₁₂
- Description:Benz(a)anthracene
- CAS No.:56-55-3



