Anthracene
Anthracene, 1,000 µg/ml in acetone, 1 ml/ampoule, 1 mL
Catalog # REST31269
Supplier: Restek
Jump to:
Product Details & Documents
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifications
Specifications
- Concentration:1,000 µg/ml in acetone
- Pk:1 mL
- Description:Anthracene
- Pk Info:1 ml/ampoule
- Cat. No.:REST31269
- Formula:C₁₄H₁₀
- Description:Anthracene
- CAS No.:120-12-7



