Atrazine
Atrazine, 1,000 µg/ml in acetone, 1 ml/ampoule, 1
Catalog # REST32208
Supplier: Restek
Jump to:
Product Details & Documents
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifications
Specifications
- Concentration:1,000 µg/ml in acetone
- Pk:1
- Description:Atrazine
- Pk Info:1 ml/ampoule
- Cat. No.:REST32208
- Formula:C₈H₁₄ClN₅
- Description:Atrazine
- CAS No.:1912-24-9



