Organic reference standards, Carbazole
Supplier: Restek
About this item
: Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis and material safety data sheet.
2 Options Available Below
Product Details & Documents
: Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis and material safety data sheet.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- REST31430
- REST31836
- Description:
- Carbazole
- Carbazole
- CAS No.:
- 86-74-8
- 86-74-8
- Formula:
- C₁₂H₉N
- C₁₂H₉N
- Melting point:
- 243…246 °C (lit.)
- 243…246 °C (lit.)
Specifications
Product Family Options
Product Information
- ConcentrationPk InfoPkAvailabilityPrice
- REST31430CarbazoleSpecifications:
More
Specifications
Cat. No.REST31430DescriptionCarbazoleCAS No.86-74-8FormulaC₁₂H₉NMelting point243…246 °C (lit.) - REST31836CarbazoleSpecifications:
More
Specifications
Cat. No.REST31836DescriptionCarbazoleCAS No.86-74-8FormulaC₁₂H₉NMelting point243…246 °C (lit.)



