Trichlorofluoromethane (CFC-11)
Catalog # REST30421
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml in P&T methanol
- Pk:1 mL
- Standards:CRM
- CAS No.:75-69-4
- Description:Trichlorofluoromethane (CFC-11)
- Formula:CFCl₃
- Description:Trichlorofluoromethane (CFC-11)
- Pk Info:1 ml/ampoule
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.