Orders LinkContactUsLinkComponent
Isocyanates singles
Supplier:   Restek
Isocyanates singles
Supplier:  Restek
Certificates
Isocyanates singles
Isocyanates singles
4 Options Available Below
Product Details & Documents
OSHA 47 is for 4,4'-methylenediphenyl isocyanate (4,4'-MDI) in indoor air, and NIOSH Method 5522 is for 2,4-TDI; 2,6-TDI; 4,4'-MDI; and 1,6-hexamethylene diisocyanate (1,6-HDI) in air. Restek offers the 1,-(2-pyridyl)piperzine (1-2pp) derivative.
ASTM D5836 and OSHA 42 are test methods for determining 2,4-toluene diisocyanate (2,4-TDI) and 2,6-TDI in the workplace atmosphere.
Product Family Options
Product Information
  • Pk Info
    Pk
    Availability
    Price
  • REST33000
    2,6-TDIP, 1000 µg/ml in dimethyl sulfoxide
    2,6-TDIP, 1000 µg/ml in dimethyl sulfoxide
    1 ml/ampoule
    1 Ampoul
    Log in for availability
    Log in for availability
    1 * 1 Ampoul
    1 * 1 Ampoul - 60,50 €
    60,50 €
    1 * 1 Ampoul
       60,50 €
  • REST33001
    2,4-TDIP,1000 µg/ml in dimethyl sulfoxide
    2,4-TDIP,1000 µg/ml in dimethyl sulfoxide
    1 ml/ampoule
    1 Ampoul
    Log in for availability
    Log in for availability
    1 * 1 Ampoul
    1 * 1 Ampoul - 60,50 €
    60,50 €
    1 * 1 Ampoul
       60,50 €
  • REST33002
    1,6-HDIP, 1000 µg/ml in dimethyl sulfoxide
    1,6-HDIP, 1000 µg/ml in dimethyl sulfoxide
    1 ml/ampoule
    1 Ampoul
    Log in for availability
    Log in for availability
    1 * 1 Ampoul
    1 * 1 Ampoul - 60,50 €
    60,50 €
    1 * 1 Ampoul
       60,50 €
  • REST33003
    4,4'-MDIP, 1000 µg/ml in dimethyl sulfoxide
    4,4'-MDIP, 1000 µg/ml in dimethyl sulfoxide
    1 ml/ampoule
    1 Ampoul
    Log in for availability
    Log in for availability
    1 * 1 Ampoul
    1 * 1 Ampoul - 59,50 €
    59,50 €
    1 * 1 Ampoul
       59,50 €