ContactUsLinkComponent
FAME #12 mix
Catalog # REST35021
Supplier:  Restek
CAS Number:  
undefined
FAME #12 mix
Catalog # REST35021
Supplier:  Restek
CAS Number:  
Specifications
  • Pack type:
    Ampoule
  • Pk:
    1
  • Description:
    FAME #1, each mixture has minimum 50 mg/ampoule
Specifications
About this item
This mixture can be used for quantification (AOCS method CE 1a-13) and approximate the compositions of oils.
  • Suitable for oils of mid-range to long chain lengths (C13 - C21)
This mix contains five components such as C13 Methyl tridecanoate (20%), C15 methyl pentadecanoate (20%), C17 methyl heptadecanoate (20%), C19 methyl nonadecanoate (20%), C21 methyl heneicosanoate (20%).