ContactUsLinkComponent
FAME #9 mix
FAME #9 mix
  REST35018
 :  Restek
 :  
undefined
FAME #9 mix
  REST35018
 :  Restek
 :  
  • Pack type:
    Ampoule
  • Pk:
    1
  • Description:
    FAME #9, each mixture has minimum 50 mg/ampoule
This mixture can be used for quantification (AOCS method CE 1a–13) and approximate the compositions of oils.
  • Suitable for oils of mid-range to long chain lengths (C16 - C20)
This mix contains five components such as C16:00 methyl palmitate (20%), C17:00 methyl heptadecanoate (20%), C18:00 methyl stearate (20%), C19:00 methyl nonadecanoate (20%), C20:00 methyl arachidate (20%).