Teclu safety burner
Supplier: JUCHHEIM Laborgerate GmbH
Certificates
About this item
Teclu safety burner with needle valve and thermofeeler with safety valve on heavy, round base.
Height: 200 mm
Weight: 685 g
Teclu safety burner with needle valve and thermofeeler with safety valve on heavy, round base.
Height: 200 mm
Weight: 685 g