Ethylene oxide standard
Ethylene oxide standard, 0,005% w/w in PEG-200, 5 ml/ampoule, 1 Ampoul
Supplier: Restek
Jump to:
Product Details & Documents
Used as reference standards for detecting ethylene oxide and 1,4-dioxane in PEG-based products.
- Formulated with concentrations needed by labs analysing cosmetics and personal care products for possible carcinogens
- Quantitatively tested to confirm composition; detailed support documentation provided
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:1 Ampoul
- Description:Ethylene oxide standard, 0,005% w/w in PEG-200
- Pk Info:5 ml/ampoule
- Cat. No.:REST30548
- Formula:C₂H₄O
- Description:Ethylene oxide
- CAS No.:75-21-8



