Acifluorfen, reference standards
REST32255
: Restek
:
- Pk:1
- CAS No.:50594-66-6
- Description:Acifluorfen, 1000 µg/ml in methanol
- Formula:C₁₄H₇ClF₃NO₅
- Description:Acifluorfen
- Pk Info:1 ml/ampoule
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.